Order Entry
Sweden
ContactUsLinkComponent
(R)-(-)-2-Phenylbutyric acid 99%
Irritant
(R)-(-)-2-Phenylbutyric acid 99%
Certificates
undefined
Irritant
(R)-(-)-2-Phenylbutyric acid 99%

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5150 to 1.5170 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information