L(+)-Valine ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:72-18-4
:
Formula:C₅H₁₁NO₂
MDL Number:MFCD00064220
:117.15 g/mol
Merck Index:14,9909
Boiling Pt:217 °C (1013 hPa)
Melting Pt:∼ 315 °C
Density:1.23
Storage Temperature:Ambient
EINECS:200-773-6