Order Entry
Sweden
ContactUsLinkComponent
L(+)-Tartaric acid, powder, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, E334, SAFC®
Corrosive
L(+)-Tartaric acid, powder, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, E334, SAFC®
Catalog # 1.00803.9029
CAS Number:  
undefined
Corrosive
L(+)-Tartaric acid, powder, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, E334, SAFC®
Catalog # 1.00803.9029
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
Merck Index:13,09158
Boiling Pt:399 °C (1013 hPa)
Melting Pt:168…170 °C
Density:1.759 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Flash Pt:150 °C (closed cup)
 :150.09 g/mol
Storage Temperature:Ambient
EINECS:201-766-0
REACH:01-2119537204-47
 :GHS05
CAS Number:87-69-4
Formula:C₄H₆O₆
MDL Number:MFCD00064207