D-(-)-Aspartic acid
Glass bottle for solids, 25 g
Catalog # MOLE12844702-25G
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH
- CAS Number:1783-96-6
- Formula:C₄H₇NO₄
- MDL Number:MFCD00063081
- Molecular Weight:133.1 g/mol
- Merck Index:13,00845
- Melting Pt:251 °C
- EINECS:217-234-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



