Organic reference standard, 2,2',3,5,5',6-Hexachlorobiphenyl (PCB No. 151) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-151
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:52663-63-5
- Description:2,2',3,5,5',6-Hexachlorobiphenyl (PCB No. 151) 35 µg/ml in isooctane
- Formula:C₁₂H₄Cl₆
- Description:2,2',3,5,5',6-Hexachlorobiphenyl (PCB No. 151)