Order Entry
Sweden
ContactUsLinkComponent
Iron(II) sulphate heptahydrate ≥96%, TECHNICAL
Iron(II) sulphate heptahydrate ≥96%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Iron(II) sulphate heptahydrate ≥96%, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 96 %

Specification Test Results