Order Entry
Sweden
ContactUsLinkComponent
Zinc sulphate heptahydrate, BAKER, Laboratory Reagent, J.T.Baker®
Zinc sulphate heptahydrate, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 1796.1000
Leverantör:  Avantor
CAS-nummer:  
Zinc sulphate heptahydrate, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 1796.1000
Leverantör:  Avantor
CAS-nummer:  

Specifikationer

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifikationer

Specifikationstestresultat

  • Assay:
    99.0 - 104.0 %
  • Acidity:
    passes test
  • Alkalies and Alkaline Earths:
    max. 0.9 %
  • Appearance of solution:
    passes test
  • Arsenic (As):
    passes test
  • Chloride (Cl):
    max. 300 ppm
  • Iron (Fe):
    max. 100 ppm
  • Lead (Pb):
    max. 0.002 %
  • Identification:
    passes test
  • pH:
    4.4 - 5.6

Specifikationstestresultat

Om denna produkt

 :
Merck Index:12,10293
Smält Pt:100 °C
Densitet:1.97 g/cm³ (20 °C)
ADR:9,III
 :287.54 g/mol
EINECS:231-793-3
FN:3077
 :GHS05
 :GHS07
 :GHS09
Förvaringstemperatur:Ambient
CAS-nummer:7446-20-0
Formel:ZnSO₄·7H₂O
MDL-nummer:MFCD00149894