Zinc sulphate heptahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7446-20-0
:
Formula:ZnSO₄·7H₂O
MDL Number:MFCD00149894
Merck Index:14,10159
Melting Pt:100 °C
Density:1.97
Storage Temperature:Ambient
EINECS:231-793-3
UN:3077
ADR:9,III
:287.54 g/mol
:GHS05
:GHS07
:GHS09