Zinc sulphate monohydrate (35.5% Zn)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7446-19-7
:
Formula:ZnSO₄·1H₂O
MDL Number:MFCD00149893
:179.45 g/mol
Merck Index:14,10159
Density:3.2 g/cm³ (20 °C)
EINECS:231-793-3
UN:3077
ADR:9,III
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient