Order Entry
Sweden
ContactUsLinkComponent
 
 
 
 
Copper(II) sulphate pentahydrate, powder, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
 
 
 
 
Copper(II) sulphate pentahydrate, powder, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
 

 

  • Assay (iodometric):
    99.0 - 100.5 %
  • Assay (iodometric, dried substance):
    98.5 - 100.5 %
  • Identity:
    passes test
  • Appearance of solution:
    passes test
  • Chloride (Cl):
    ≤ 0.0100 %
  • Al (Aluminium):
    ≤ 0.0005 %
  • Ca (Calcium):
    ≤ 0.005 %
  • Cr (Chromium):
    ≤ 0.005 %
  • Fe (Iron) (*):
    ≤ 0.003 %
  • K (Potassium):
    ≤ 0.01 %
  • Na (Sodium):
    ≤ 0.02 %
  • Ni (Nickel) (*):
    ≤ 0.005 %
  • Pb (Lead):
    ≤ 0.0050 %
  • Zn (Zinc):
    ≤ 0.03 %
  • Residual solvents (ICH Q3C):
    excluded by manufacturing process
  • Loss on drying (250 °C):
    35.0 - 36.5 %
  • Particle size (< 0,18 mm):
    100%

 

Frequently Bought Together