


Copper(II) nitrate hemipentahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19004-19-4
:
Formula:Cu(NO₃)₂·2.5H₂O
MDL Number:MFCD02253001
:232.59 g/mol
Merck Index:14,2643
Boiling Pt:170 °C (dec.)
Melting Pt:∼115 °C
Density:2.32
Storage Temperature:Ambient
EINECS:221-838-5
UN:3085
ADR:5.1,III
:GHS03
:GHS05
:GHS07