Calcium sulphate dihydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10101-41-4
:
Formula:CaSO₄·2H₂O
MDL Number:MFCD00149625
:172.17 g/mol
Merck Index:14,1706
Boiling Pt:163 °C (–2H₂O)
Melting Pt:128 °C (-1.5H₂O)
Density:2.32
Storage Temperature:Ambient
EINECS:231-900-3