Talc
25 kg
Catalog # SWEDTA917403
Supplier: SWED HANDLING
Jump to:
Product Details & Documents
Synonyms: Magnesium silicate monohydrate, Soapstone, Talcum venetum
- CAS Number:14807-96-6
- Formula:Mg₃O₁₁Si₄·1H₂O
- MDL Number:MFCD00792903
- Molecular Weight:379.27 g/mol
- Merck Index:13,09127
- Melting Pt:>1200 °C
- Density:2.58…2.83 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:238-877-9
Specifications
Specifications
- Pk:25 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



