Order Entry
Sweden
ContactUsLinkComponent
Hydrazinium sulphate ≥99.0% ACS
Toxic
Health
Irritant
Hydrazinium sulphate ≥99.0% ACS
Catalog # 40120.36
CAS Number:  
undefined
Toxic
Health
Irritant
Hydrazinium sulphate ≥99.0% ACS
Catalog # 40120.36
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

CAS Number:10034-93-2
 :
Formula:NH₂NH₂·H₂SO₄
MDL Number:MFCD00044873
 :130.12 g/mol
Merck Index:14,4772
Melting Pt:∼ 255 °C (dec.)
Density:1.370
EINECS:233-110-4
UN:3288
ADR:6.1,III
 :GHS06
 :GHS08
 :GHS07
Storage Temperature:Ambient