Order Entry
Sweden
ContactUsLinkComponent
 
Guanidinium hydrochloride
Catalog # SHETIB05080
Supplier:  IBI SCIENTIFIC
CAS Number:  
undefined
Guanidinium hydrochloride
Catalog # SHETIB05080
Supplier:  IBI SCIENTIFIC
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :95.53 g/mol
Merck Index:13,04578
Melting Pt:185 °C
Density:1.354 g/cm³ (20 °C)
EINECS:200-002-3
 :GHS07
Storage Temperature:Ambient
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026