
Diethylamine hydrobromide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6274-12-0
:
Formula:C₄H₁₂BrN
MDL Number:MFCD00012498
:154.05 g/mol
:M-Clarity™ quality level = MQ200
EINECS:228-466-2
:GHS07
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:215 - 218 °C
- Molar Mass:154.05 g/mol