Order Entry
Sweden
Orders LinkContactUsLinkComponent
Nitric acid 53%

Nitric acid 53%

Glass bottle, 1 L
Catalog # ROTH9274.1
Corrosive
Marketsource
Supplier:   Roth Carl
Nitric acid 53%
Catalog # ROTH9274.1
Supplier:  Roth Carl
Marketsource
CAS Number:  7697-37-2
Nitric acid 53%

Glass bottle, 1 L

CAS Number:  7697-37-2
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Nitric acid solution
  • CAS Number:7697-37-2
  • Formula:HNO₃
  • MDL Number:MFCD00011349
  • Molecular Weight:63.01 g/mol
  • Storage Temperature:Ambient
  • ADR:8,II
  •  :Density: 1.51 g/cm³ (20 °C)
  • EINECS:231-714-2
  • UN:2031
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications