Order Entry
Sweden
ContactUsLinkComponent
Magnesium chloride 1 mol/l (2 N) in aqueous solution
Magnesium chloride 1 mol/l (2 N) in aqueous solution
Catalog # AMBI9530G
Supplier:  Ambion
CAS Number:  
Magnesium chloride 1 mol/l (2 N) in aqueous solution
Catalog # AMBI9530G
Supplier:  Ambion
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :95.21 g/mol
Merck Index:13,05684
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7786-30-3
Formula:MgCl₂
MDL Number:MFCD00011106