Sodium hydroxide standardised solution 0.1 N in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III