Order Entry
Sweden
ContactUsLinkComponent
(±)-1-Phenoxy-2-propanol
(±)-1-Phenoxy-2-propanol
Catalog # EHERC16045600
Supplier:  EHRENSTORFER
CAS Number:  
(±)-1-Phenoxy-2-propanol
Catalog # EHERC16045600
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
Boiling Pt:243 °C (1013 hPa)
Melting Pt:11.4 °C
Density:1.0665 g/cm³ (20 °C)
 :152.19 g/mol
Storage Temperature:Ambient
EINECS:212-222-7
CAS Number:770-35-4
Formula:C₆H₅OCH₂CH(OH)CH₃
MDL Number:MFCD00016861