Polysorbate 60
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9005-67-8
:
Formula:C₆₄H₁₂₆O₂₆
MDL Number:MFCD01780110
Boiling Pt:? 110 °C (1013 hPa)
Density:1.08 g/cm³
Flash Pt:>150 °C
Storage Temperature:Ambient
EINECS:500-020-4
Specification Test Results
- Appearance:Light yellow to light brown Paste
- Infrared spectrum:Conforms
- Water:≤3 %
- Dioxane:≤5 ppm
- Acid value:≤2 mg KOH/g
- Hydroxyl value:81 to 96 mg KOH/g
- Saponification value:45 to 55 mg KOH/g