Order Entry
Sweden
ContactUsLinkComponent
Nitrile gloves, Dexpure® 800-91
Nitrile gloves, Dexpure® 800-91
Supplier:  HONEYWELL SAFETY
Certificates
undefined
Nitrile gloves, Dexpure® 800-91
Supplier:  HONEYWELL SAFETY
Specifications
  • Catalog No:
  • DALL4580091/L
  • DALL4580091/M
  • DALL4580091/S
  • DALL4580091/XL
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Low-powdered
  • Low-powdered
  • Low-powdered
  • Low-powdered
  • Finish:
  • Textured
  • Textured
  • Textured
  • Textured
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • EN norm:
  • EN ISO 374-1:2016 Type C, EN ISO 374-5:2016 (MICRO-ORGANISMS)
  • EN ISO 374-1:2016 Type C, EN ISO 374-5:2016 (MICRO-ORGANISMS)
  • EN ISO 374-1:2016 Type C, EN ISO 374-5:2016 (MICRO-ORGANISMS)
  • EN ISO 374-1:2016 Type C, EN ISO 374-5:2016 (MICRO-ORGANISMS)
  • Food contact approved:
  • +
  • +
  • +
  • +
Specifications