Order Entry
Sweden
ContactUsLinkComponent
Tween® 20 (Polysorbate)
Tween® 20 (Polysorbate)
Catalog # RESERES9691T-A101X
Supplier:  RESEARCH ORGANICS
CAS Number:  
Tween® 20 (Polysorbate)
Catalog # RESERES9691T-A101X
Supplier:  RESEARCH ORGANICS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Seq:
    2440

Specifications

About this item

 :
 :1227.53 g/mol
Density:1.1 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:500-018-3
CAS Number:9005-64-5
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986