Polysorbate 20, Reagent Grade
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:9005-64-5
:
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986
Boiling Pt:>100 °C
Density:1.1 g/cm³ (20 °C)
Flash Pt:149 °C (closed cup)
Storage Temperature:Ambient
EINECS:500-018-3
Specification Test Results
- Arsenic:<0.0003%
- DNase:NONE
- Heavy Metals:<0.001%
- Hydroxyl Number:96 – 108
- Identification:PASS
- Moisture (KF):3.0%
- Protease:NONE
- Residue on Ignition:0.25%
- RNase:NONE