Order Entry
Sweden
ContactUsLinkComponent
Paraformaldehyde for synthesis, Sigma-Aldrich®
Corrosive
Health
Irritant
Paraformaldehyde for synthesis, Sigma-Aldrich®
Catalog # 8.18715.9025
Supplier:  Merck
CAS Number:  
Corrosive
Health
Irritant
Paraformaldehyde for synthesis, Sigma-Aldrich®
Catalog # 8.18715.9025
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    100 - 130 °C
  • Vapor Pressure:
    1.93 hPa (25 °C)
  • Flash Point:
    70 °C
  • Explosion Limit:
    7 - 73 %(V)
  • Density:
    1.4 g/cm3 (20 °C)
  • pH:
    5.5 (H2O, 20 °C) (saturated solution)
  • Ignition Point:
    300 °C
  • Hazard Class:
    4.1
  • Hazard Class Description:
    Flammable Solid

Specification Test Results

About this item

 :
Merck Index:13,07096
Boiling Pt:135…136 °C (13 mmHg)
Melting Pt:123 °C
Density:1.46 g/cm³ (20 °C)
Flash Pt:70 °C
Storage Temperature:Refrigerator
UN:2213
ADR:4.1,III
 :M-Clarity™ quality level = MQ200
 :GHS05
 :GHS08
 :GHS07
CAS Number:30525-89-4
Formula:(CH₂O)n
MDL Number:MFCD00133991