Order Entry
Sweden
ContactUsLinkComponent
Polyethylene glycol 1,000, Supelco®
Polyethylene glycol 1,000, Supelco®
Catalog # 81288-1G
Supplier:  Merck
CAS Number:  
undefined
Polyethylene glycol 1,000, Supelco®
Catalog # 81288-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :1000 g/mol
Merck Index:13,07651
Boiling Pt:∼250 °C (1013 hPa)
Melting Pt:33…40 °C
Density:1.2 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Flash Pt:260 °C
Storage Temperature:Ambient
EINECS:500-038-2
CAS Number:25322-68-3
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839