Polyethylene glycol 1,000
Catalog # ROTH0150.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:1000 g/mol
Merck Index:13,07651
Boiling Pt:∼250 °C (1013 hPa)
Melting Pt:33…40 °C
Density:1.2 g/cm³ (20 °C)
Flash Pt:260 °C
Storage Temperature:Ambient
EINECS:500-038-2
CAS Number:25322-68-3
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839