Order Entry
Sweden
ContactUsLinkComponent
Gum arabic
Gum arabic
Catalog # ROTH4159.1
Supplier:  Roth Carl
CAS Number:  
Gum arabic
Catalog # ROTH4159.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

About this item

 :
Density:1,35 g/cm³ (20 °C)
Flash Pt:>250 °C
EINECS:232-519-5
 :GHS07
Storage Temperature:Ambient
CAS Number:9000-01-5
MDL Number:MFCD00081264