
Gum arabic
Catalog # MOLE13599707-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Density:1,35 g/cm³ (20 °C)
Flash Pt:>250 °C
EINECS:232-519-5
:GHS07
Storage Temperature:Ambient
CAS Number:9000-01-5
MDL Number:MFCD00081264