Gum arabic, powder
:9000-01-5
:
:MFCD00081264
:1.35 g/cm³
:≥ 250 °C
:232-519-5
:GHS07
:Ambient
- Appearance:White to yellow to light beige Powder
- Infrared spectrum:Conforms
- Loss on drying:≤15.0 % (1 g, 105°C)
- Additional info:Agar, Starch, Tragacanth, dextrin, sucrose: absent
- Identification:Passes test (per Ph. Eur.)
- Total ash:≤4.0 %
- Arsenic (As):≤3 ppm
- Lead (Pb):≤10 ppm
- Heavy metals (as Pb):≤40 ppm
- Insoluble matter:≤0.1 %