Order Entry
Sweden
ContactUsLinkComponent
Agarose
Agarose
Catalog # MOLEM76055259
Supplier:  Molekula
CAS Number:  
undefined
Agarose
Catalog # MOLEM76055259
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
Melting Pt:60…90 °C
Storage Temperature:Ambient
EINECS:232-731-8
CAS Number:9012-36-6
MDL Number:MFCD00081294