Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(+)-Maltose monohydrate
D-(+)-Maltose monohydrate
D-(+)-Maltose monohydrate
D-(+)-Maltose monohydrate
MOLEM19997254
:
Molekula
:
D-(+)-Maltose monohydrate
MOLEM19997254
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
1 kg
Seq:
4024
:
:
360.32 g/mol
:
13,05736
:
160…165 °C
:
Ambient
:
200-716-5
:
6363-53-7
:
C₁₂H₂₂O₁₁·1H₂O
:
MFCD00149343