Om denna produkt
Designed for ease of use and safe operation, the shiroGEL horizontal electrophoresis systems and accessories are built to withstand the rigours of everyday use. For leakproof performance the gel boxes and gel trays are moulded from thick acrylic. UV transparent gel trays aid in visualisation of samples. Cassettes protect the electrodes and allow for easy replacement. Gel casting is simple with the durable rubber casting gates. Slots in the sides of the trays allow for easy comb placement. The two-piece design of the combs allows for vertical adjustment, giving the user control over the depth of the well. The combs supplied with the systems are 1,5 mm thick. Multiple combs, including those compatible with multichannel pipettes and different thicknesses, are also available. The 'EasyLift' gel box lid is easily removed using the side tabs and pressure pads. The lid is domed, to prevent condensation from dripping onto the gel, and is surrounded by a drip ring, to help recover the condensate and maintain buffer concentration.
- Moulded, durable construction
- Rubber gates for tapeless gel casting
- 'EasyLift' safety lid with buffer recapture system to maintain buffer concentration
*Multichannel pipette compatible
Specifikationer
- Catalog No:
- 700-0255
- 700-0253
- 700-0250
- Model:
- Midi Plus 15
- Midi Plus 10
- Mini 10
- Gel size W×L:
- 150×150 mm
- 150×100 mm
- 70×100 mm
- Sample capacity:
- 210
- 140
- 64
- Buffer volume:
- 500 ml
- 300 ml
- 225 ml
- Unit W×L×H:
- 175×265×90 mm
- 175×265×90 mm
- 90×210×90 mm