Molecular sieve 13X (1.0 nm, 10 Å), pellets, Ø 3.2 mm, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:63231-69-6
:1318-02-1
MDL Number:MFCD00131613
Storage Temperature:Ambient
Specification Test Results
- Particle Size:3.2 mm
- Form:pellets
- Granulation:complying
- Pore Size (Ång Ström):10