Order Entry
Sweden
ContactUsLinkComponent
 
Molecular sieve 13X (1.0 nm, 10 Å), powder ∼2 μm, Fluka™
  283592-5KG
 :  
Molecular sieve 13X (1.0 nm, 10 Å), powder ∼2 μm, Fluka™
  283592-5KG
 :  

 

  • Pack type:
    Drum with inliner
  • Pk:
    5 kg
  • Seq:
    4114

 

 

  • Loss on ignition (575°C):
    Max. 1 %
  • Water absorbing capacity:
    Min. 20 %

 

 

 :
 :Ambient
 :63231-69-6
 :1318-02-1
 :MFCD00131613