Paraffin, Solidification point: 52-54°C
Catalog # SWED13645-20
Supplier: SWED HANDLING
CAS Number:
Specifications
- Pk:20 kg
- Seq:4241
Specifications
About this item
:
Density:0.9 g/cm³ (20 °C)
EINECS:232-315-6
CAS Number:8002-74-2
:64742-51-4