Order Entry
Sweden
ContactUsLinkComponent
Organic reference standards, dicloxacillin sodium salt hydrate, VETRANAL™, Supelco®
Organic reference standards, dicloxacillin sodium salt hydrate, VETRANAL™, Supelco®
  46182-100MG
 :  Merck
 :  
undefined
Organic reference standards, dicloxacillin sodium salt hydrate, VETRANAL™, Supelco®
  46182-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    13412-64-1
  • Description:
    Dicloxacillin sodium salt hydrate
  • M-Clarity quality level:
    MQ200
  • Formula:
    C₁₉H₁₆Cl₂N₃NaO₅S·xH₂O
  • Storage temperature:
    2...8 °C
  • Description:
    Dicloxacillin sodium salt hydrate

 

 

Suitable for HPLC and Gas chromatography (GC) applications.