Order Entry
Sweden
ContactUsLinkComponent
Tin, foil, Thickness: ∼ 0.04 mm, Supelco®
Tin, foil, Thickness: ∼ 0.04 mm, Supelco®
Catalog # 1.07826.0001
Supplier:  Merck
CAS Number:  
Tin, foil, Thickness: ∼ 0.04 mm, Supelco®
Catalog # 1.07826.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic box
  • Pk:
    200
  • Seq:
    0200

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    232 °C
  • Molar Mass:
    118.71 g/mol
  • Boiling Point:
    2260 °C (1013 hPa)
  • Density:
    7.3 g/cm3 (20 °C)

Specification Test Results

About this item

 :
Merck Index:12,09587
Boiling Pt:2270 °C (1013 hPa)
Melting Pt:232 °C
Density:7.31 g/cm³ (25 °C)
 :118.71 g/mol
Storage Temperature:Ambient
EINECS:231-141-8
CAS Number:7440-31-5
Formula:Sn
MDL Number:MFCD00133862