Order Entry
Sweden
ContactUsLinkComponent
Tin ≥99.999% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Tin ≥99.999% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Catalog # 38538.FY
CAS Number:  
Tin ≥99.999% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Catalog # 38538.FY
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    1724
  • Pk Info:
    50×100 mm

Specifications

About this item

 :
Merck Index:12,09587
Boiling Pt:2270 °C (1013 hPa)
Melting Pt:232 °C
Density:7.31 g/cm³ (25 °C)
 :118.71 g/mol
Storage Temperature:Ambient
EINECS:231-141-8
CAS Number:7440-31-5
Formula:Sn
MDL Number:MFCD00133862