Tin ≥99.99% (metals basis), shot 3 mm (0.1 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-31-5
:
Formula:Sn
MDL Number:MFCD00133862
Merck Index:12,09587
Boiling Pt:2270 °C (1013 hPa)
Melting Pt:232 °C
Density:7.31 g/cm³ (25 °C)
:118.71 g/mol
Storage Temperature:Ambient
EINECS:231-141-8