0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tin
Tin ≥99.9985% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)
Tin ≥99.9985% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)
:
Thermo Fisher Scientific
Tin ≥99.9985% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)
:
Thermo Fisher Scientific
:
7440-31-5
:
:
Sn
:
MFCD00133862
:
12,09587
:
2270 °C (1013 hPa)
:
232 °C
:
7.31 g/cm³ (25 °C)
:
118.71 g/mol
:
Ambient
:
231-141-8
externalurl.pdf
msds_ALFA011457