Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tenn
Tenn ≥99.85%, pulver -100 mesh
Tenn ≥99.85%, pulver -100 mesh
Tenn ≥99.85%, pulver -100 mesh
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Tenn ≥99.85%, pulver -100 mesh
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
7440-31-5
:
Formula:
Sn
MDL Number:
MFCD00133862
Storage Temperature:
Omgivande
ADR:
4.1,III
:
118.71 g/mol
EINECS:
231-141-8
UN:
3089
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA000941
Videor
Thermo Scientific Acroseal 25 Years
Documentation