Order Entry
Sweden
ContactUsLinkComponent
Tin ACS 20 mesh
Tin ACS 20 mesh
Certificates
undefined
Tin ACS 20 mesh

Specification Test Results

  • Appearance:
    Grey Metallic granular powder
  • Titration Complexometric:
    ≥99.5 %
  • Antimony (Sb):
    ≤200 ppm
  • Arsenic (As):
    ≤1 ppm
  • Copper (Cu):
    ≤50 ppm
  • Iron (Fe):
    ≤100 ppm
  • Lead (Pb):
    ≤50 ppm

Specification Test Results

Related Information