Gå till Rubrik
Gå till Huvudinnehåll
Gå till Sidfot
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Graphite
Graphite (synthetic) ≥99.9995% (metals basis), powder -325 mesh
Graphite (synthetic) ≥99.9995% (metals basis), powder -325 mesh
:
Thermo Fisher Scientific
Graphite (synthetic) ≥99.9995% (metals basis), powder -325 mesh
:
Thermo Fisher Scientific
:
7782-42-5
:
:
C
:
MFCD00144065
:
12,04560
:
3827 °C
:
2.09…2.23 g/cm³ (20 °C)
:
231-955-3
:
1325
:
4.1,III
:
12.01 g/mol
:
GHS02
:
GHS07
:
Ambient
externalurl.pdf
msds_ALFA040798
Thermo Scientific Acroseal 25 Years