Graphite ≥99.9995% (metals basis), rods, Length: 152mm (6in), Ø 6.15mm (0.242in)
Catalog # 40767.KD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50
- Seq:0050
Specifications
About this item
:
Merck Index:12,04560
Melting Pt:3827 °C
Density:2.09…2.23 g/cm³ (20 °C)
:12.01 g/mol
Storage Temperature:Ambient
EINECS:231-955-3
CAS Number:7782-42-5
Formula:C
MDL Number:MFCD00144065