Organic reference standard, Phthalic acid bis-isononyl ester, Dr Ehrenstorfer
Catalog # EHERC16173600
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:0,25 g
- Standards:Reference standard
- CAS No.:68515-48-0
- Description:Phthalic acid, bis-isononyl ester
- Formula:C₁₀H₁₀O₄(CH₂)n(CH₂)n
- Description:Phthalic acid, bis-isononyl ester
- Molecular weight (Mw):222,24 g/mol
Specifications
About this item
Certifications: Meets ISO/IEC 17025 specifications.