Order Entry
Sweden
ContactUsLinkComponent
Sodium hypochlorite solution 5% (w/w), Technical Grade
Sodium hypochlorite solution 5% (w/w), Technical Grade
Supplier:  PanReac AppliChem
Certificates
Sodium hypochlorite solution 5% (w/w), Technical Grade
Supplier:  PanReac AppliChem
Certificates

Specification Test Results

  • Assay (iodometr.):
    5% ± 0.5% (At the moment of the batch analysis)

Specification Test Results

Related Information