Storage cabinet
Catalog # SEMA5187
Supplier: SEMADENI
Specifications
- Description:Storage system, light grey, PS
- W×D×H:80×480×45 mm
- Pk:1
Specifications
About this item
The storage cabinet system allows a quick, individual and efficient division of drawers, shelves, cupboards, etc.
- The boxes can be put together in a number of variations according to a sophisticated module
- The boxes have holders on the bottom, which fit into the grid area of the self-adhesive drawer foil
- This self-adhesive foil prevents the boxes from slipping around in the drawer
- Heavy-duty boxes made of impact-resistant polystyrene (PS).