Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG Human Polyclonal Antibody (Unconjugated)
Anti-IgG Human Polyclonal Antibody (Unconjugated)
Catalog # IMMRIR2368
Supplier:  ImmunoReagents
Anti-IgG Human Polyclonal Antibody (Unconjugated)
Catalog # IMMRIR2368
Supplier:  ImmunoReagents

Specifications

  • Antibody type:
    Control
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Human
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Form:
    Liquid
  • Storage temperature:
    Cool
  • Concentration:
    > 1.0 mg/ml (E 1% at 280 nm = 13.0)
  • Shipping temperature:
    2-8 °C
  • Size:
    2 mg
  • Pk:
    2 mg

Specifications

About this item

Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype:
Reactivity: