Anti-IgG Rabbit Polyclonal Antibody (Unconjugated)
IMMRRBXSH-003-D
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Sheep
- Form:Clear, colorless liquid, 0.2 µm filtered
- Concentration:>4.5 mg/ml (E 1% at 280nm=13.0)
- Immunogen:Purified Sheep IgG, whole molecule
- Purification:Affinity purified using solid phase Sheep IgG
- Pack type:Glass vial
- Pk:2 mg
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Sheep